Order Entry
Norway
ContactUsLinkComponent
1-Amino-2-methyl-9,10-anthraquinone ≥90.0% (by total nitrogen basis)
1-Amino-2-methyl-9,10-anthraquinone ≥90.0% (by total nitrogen basis)
Catalog # TCIAA1255-25G
Supplier:  TCI
CAS Number:  
undefined
1-Amino-2-methyl-9,10-anthraquinone ≥90.0% (by total nitrogen basis)
Catalog # TCIAA1255-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (HPLC):
    min. 85.0 area%
  • Purity (with Total Nitrogen):
    min. 90.0 %
  • Melting point:
    198.0 to 204.0 deg-C

Specification Test Results

About this item

 :
EINECS:201-408-3
CAS Number:82-28-0
Formula:C₁₅H₁₁NO₂
MDL Number:MFCD00001220