Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tolbutamide
Tolbutamide, Supelco®
Tolbutamide, Supelco®
Tolbutamide, Supelco®
Supplier:
Merck
Certificates
Expand image
Tolbutamide, Supelco®
Supplier:
Merck
About this item
CAS Number:
64-77-7
:
Formula:
C₁₂H₁₈N₂O₃S
MDL Number:
MFCD00027169
:
270.35 g/mol
EINECS:
200-594-3