
Methyl-4-formylbenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1571-08-0
:
Formula:C₉H₈O₃
MDL Number:MFCD00006950
:164.16 g/mol
Boiling Pt:135°C
EINECS:216-385-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:58.0 to 63.0 deg-C
- Solubility in Methanol:almost transparency