
Methyl-4-formylbenzoate 99%
:1571-08-0
:
:C₉H₈O₃
:MFCD00006950
:164.16 g/mol
:57…64 °C
:144 °C
:216-385-5
:GHS07
:Ambient
- Appearance:White to cream to light green Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:59 to 63 °C
- GC:≥98.5 %