2',5'-Dimethoxyacetanilide ≥98.0% (by GC)
Catalog # TCIAD1671-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:89.0 to 93.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
EINECS:222-423-1
CAS Number:3467-59-2
Formula:C₁₀H₁₃NO₃
MDL Number:MFCD00043826