Methyl-3-amino-5,6-dichloropyrazine-2-carboxylate 97%, Sigma-Aldrich®
Catalog # 277878-5G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance (Color):Faint Brown to Dark Brown
- Appearance (Form):Powder or Crystal or Chunk(s)
- Infrared spectrum :Conforms to Structure
- Titration by AgNO3:96.5 - 103.5 %
- Purity (HPLC):≥96.5 %
Specification Test Results
About this item
:
:222.03 g/mol
Melting Pt:230...233 °C (lit.)
:M-Clarity™ quality level = MQ200
EINECS:215-947-7
:GHS07
Storage Temperature:Room Temperature
CAS Number:1458-18-0
Formula:C₆H₅Cl₂N₃O₂
MDL Number:MFCD00010431