Tricaine methanesulphonate ≥97.0% (by HPLC, total nitrogen)
Catalog # TCIAT0941-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:145.0 to 153.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:261.29 g/mol
Merck Index:14,09621
EINECS:212-956-8
:GHS07
CAS Number:886-86-2
Formula:C₁₀H₁₅NO₅S
MDL Number:MFCD00013176