kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Tricaine methanesulphonate
Tricaine methanesulphonate, Supelco®
Tricaine methanesulphonate, Supelco®
Tricaine methanesulphonate, Supelco®
Supplier:
Merck
Certificates
Expand image
Tricaine methanesulphonate, Supelco®
Supplier:
Merck
About this item
CAS Number:
886-86-2
:
Formula:
C₁₀H₁₅NO₅S
MDL Number:
MFCD00013176
:
261.29 g/mol
EINECS:
212-956-8