Tricaine methanesulphonate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:886-86-2
:
Formula:C₁₀H₁₅NO₅S
MDL Number:MFCD00013176
:261.29 g/mol
Melting Pt:146…150 °C
EINECS:212-956-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:146 to 150 °C
- Solubility:(5% in Water) Clear solution
- Titration with NaOH:≥97.5 %
- Iron (Fe):≤5 ppm
- Magnesium (Mg):≤0.005 %
- Sodium (Na):≤0.02 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.01 %
- Nitrogen compounds (as N):≤5 ppm