Order Entry
Norway
ContactUsLinkComponent
 
Fluorescein diacetate ≥98.0% (by HPLC)
  TCIAF0240-5G
 :  TCI
 :  
undefined
Fluorescein diacetate ≥98.0% (by HPLC)
  TCIAF0240-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (HPLC):
    min. 98.0 area%
  • Melting point:
    204.0 to 210.0 deg-C
  • Solubility in Chloroform:
    almost transparency

 

 

 :
 :416.38 g/mol
 :209-877-6
 :596-09-8
 :C₂₄H₁₆O₇
 :MFCD00005062