Order Entry
Norway
ContactUsLinkComponent
 
Fluorescein diacetate cell viability stain, Sigma-Aldrich®
 
undefined
Fluorescein diacetate cell viability stain, Sigma-Aldrich®

 

  • Appearance (Colour):
    White to Faint Yellow
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Solubility (Turbidity), c = 25 mg/ml; Acetone:
    Clear
  • Solubility (Colour):
    Colourless to Faint Yellow
  • Emission Wavelength, In 0.1 N Sodium Hydroxide:
    488.0 - 494.0 nm
  • EMM:
    ≥86.000
  • Carbon:
    68.5 - 69.9 %