Fluorescein diacetate, Sigma-Aldrich®
Catalog # 343209-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:416.38 g/mol
EINECS:209-877-6
CAS Number:596-09-8
Formula:C₂₄H₁₆O₇
MDL Number:MFCD00005062