Order Entry
Norway
ContactUsLinkComponent
Estrone
Estrone
Katalog # PROOMM0282.00
CAS Number:  
Estrone
Katalog # PROOMM0282.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
EINECS:200-164-5
CAS Number:53-16-7
Formula:C₁₈H₂₂O₂
MDL Number:MFCD00003620