
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) cis- and trans-mixture ≥90.0% (by GC)
Catalog # TCIAE0798-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 90.0 % (total of isomer)
Specification Test Results
About this item
:
Boiling Pt:152°C
:192.21 g/mol
EINECS:204-467-3
:GHS07
Storage Temperature:Refrigerator
CAS Number:121-39-1
Formula:C₁₁H₁₂O₃
MDL Number:MFCD00005123