Order Entry
Norway
ContactUsLinkComponent
Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)
Irritant
Health
Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAM0042-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
Manganese (II) acetylacetonate dihydrate ≥97.0% (by titrimetric analysis)
Catalog # TCIAM0042-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Amber powder to crystal
  • Purity (Chelometric Titration):
    min. 97.0 % (calcd.on anh.substance)
  • Solubility in Methanol:
    very faint turbidity
  • Water:
    9.5 to 14.0 %

Specification Test Results

About this item

 :
EINECS:237-858-2
 :GHS07
 :GHS08
CAS Number:14024-58-9
Formula:C₁₀H₁₄MnO₄·2H₂O
MDL Number:MFCD00000022