Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Metal half-cells
Metal half-cells
Metal half-cells
Supplier:
Mettler - Toledo
Certificates
Expand image
Metal half-cells
Supplier:
Mettler - Toledo
About this item
Metal half-cells for Redox/ORP applications.
S7 sensor head offers flexibility and can easily be connected to any meter