Immobilon® Blotting Filter Papers, Chromatography Grade
Supplier: Merck
Certificates
About this item
Chromotography-grade blotting paper for western and dot blotting.
- Plain/white filter
- Hydrophilic
- Ambient shipping
For use with Amido black, CPTS, Colloidal gold, India ink and Ponceau-S red.
Chemiluminescent, colorimetric and fluorometric detection.
Specifications
- Catalog No:
- MILFIBFP0785C
- MILFIBFP0813C
- Thickness:
- 550 μm
- 550 μm