Anti-RagD Rabbit Polyclonal Antibody
CSTA4470S
- Type antilichaam:Primair
- antigeen symbool:RagD
- Clonality:Polyklonaal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Sequentie:CGLADYGDGPDSSDADPDSGTE
- Opslagtemperatuur:−20 °C
- Shipping temperature:Ambient
- Maat:100 µl
- VPE:100 µl
Type: Primary Antibodies
Antigen: RagD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human