Order Entry
Netherlands
ContactUsLinkComponent
5 ampoules de 1 mL ,,4 components: 1mg/l each of Benzo(a)anthracene [CAS:56-55-3] Chrysene [CAS:218-01-9] Benzo(b)fluoranthene [CAS:205-99-2] Benzo(a)pyrene [CAS:50-32-8] in Dichloromethane ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 mon 1 * 5 Ampoul
Catalog # CMPLRD1209422.5
New Product
5 ampoules de 1 mL ,,4 components: 1mg/l each of Benzo(a)anthracene [CAS:56-55-3] Chrysene [CAS:218-01-9] Benzo(b)fluoranthene [CAS:205-99-2] Benzo(a)pyrene [CAS:50-32-8] in Dichloromethane ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 12 mon 1 * 5 Ampoul
Catalog # CMPLRD1209422.5