Order Entry
Netherlands
ContactUsLinkComponent
Particle size (µm): 3 ID (mm): 2.1 Length (mm): 150 USP: L1 Functional Group: C18-AQ Mode: Reversed Phase Pore size (nm): 10 Surface Area (m2/g): 350 End Capping: Yes Carbon Load (%): 13 Pressure Tolerance (MPa): 50 1 * 1 ST
Catalog # SHIM227-30765-04
Supplier:  SHIMADZU
New Product
Particle size (µm): 3 ID (mm): 2.1 Length (mm): 150 USP: L1 Functional Group: C18-AQ Mode: Reversed Phase Pore size (nm): 10 Surface Area (m2/g): 350 End Capping: Yes Carbon Load (%): 13 Pressure Tolerance (MPa): 50 1 * 1 ST
Catalog # SHIM227-30765-04
Supplier:  SHIMADZU