Order Entry
Netherlands
ContactUsLinkComponent
1 component: Chromium (VI) (Cr 6+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C PACKAGING 100 ML BOTTLE,, 1 * 100 mL
Catalog # CMPLRM002093L1
New Product
1 component: Chromium (VI) (Cr 6+) 10mg/l in Water CRM, ISO 17034 and ISO 17025,,,, Conservation 12 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C PACKAGING 100 ML BOTTLE,, 1 * 100 mL
Catalog # CMPLRM002093L1