Pressure-resistant stainless steel cartridge filled with quality mixed-bed resins Performance: 950 l/h Capacity: 2800 l at 10°GSG Connection: R 3/4 1 * 1 items
Catalog # BARN02.2800
Supplier: Thermo Fisher Scientific