Order Entry
Netherlands
ContactUsLinkComponent
Mouse Substance P ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative deter mination of mouse Substance P in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other b 1 * 48 Tests
Catalog # ANTIA1824-48
Supplier:  Antibodies.com
New Product
Mouse Substance P ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative deter mination of mouse Substance P in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other b 1 * 48 Tests
Catalog # ANTIA1824-48
Supplier:  Antibodies.com