Order Entry
Netherlands
ContactUsLinkComponent
Human MGMT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MGMT in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2215-96
Supplier:  Antibodies.com
New Product
Human MGMT ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MGMT in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2215-96
Supplier:  Antibodies.com