Order Entry
Netherlands
ContactUsLinkComponent
Human Kallikrein 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Kallikrein 4 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2487-96
Supplier:  Antibodies.com
New Product
Human Kallikrein 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Kallikrein 4 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2487-96
Supplier:  Antibodies.com