Order Entry
Netherlands
ContactUsLinkComponent
Rat Heme Oxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Heme Oxygenase 1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1786-48
Supplier:  Antibodies.com
New Product
Rat Heme Oxygenase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Heme Oxygenase 1 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1786-48
Supplier:  Antibodies.com