Order Entry
Netherlands
ContactUsLinkComponent
Human Galanin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determina tion of human Galanin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA2178-96
Supplier:  Antibodies.com
New Product
Human Galanin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determina tion of human Galanin in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 96 Tests
Catalog # ANTIA2178-96
Supplier:  Antibodies.com