Order Entry
Netherlands
ContactUsLinkComponent
Rat HSP27 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat HSP27 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2296-96
Supplier:  Antibodies.com
New Product
Rat HSP27 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat HSP27 in serum, plasma, tissue homogenates, cell culture supernates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA2296-96
Supplier:  Antibodies.com