Order Entry
Netherlands
ContactUsLinkComponent
Mouse PAPP A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PAPP A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1735-96
Supplier:  Antibodies.com
New Product
Mouse PAPP A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PAPP A in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1735-96
Supplier:  Antibodies.com