Order Entry
Netherlands
ContactUsLinkComponent
Human LIPG ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human LIPG in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1731-96
Supplier:  Antibodies.com
New Product
Human LIPG ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human LIPG in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1731-96
Supplier:  Antibodies.com