Order Entry
Netherlands
ContactUsLinkComponent
Mouse Carbohydrate Antigen 125 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse Carbohydrate Antigen 125 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1788-96
Supplier:  Antibodies.com
New Product
Mouse Carbohydrate Antigen 125 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of mouse Carbohydrate Antigen 125 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA1788-96
Supplier:  Antibodies.com