Order Entry
Netherlands
ContactUsLinkComponent
Monkey Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of Monkey Interferon gamma in serum, plasma, and cell culture supernatant. 1 * 96 Tests
Catalog # ANTIA270334-96
Supplier:  Antibodies.com
New Product
Monkey Interferon gamma ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of Monkey Interferon gamma in serum, plasma, and cell culture supernatant. 1 * 96 Tests
Catalog # ANTIA270334-96
Supplier:  Antibodies.com