Order Entry
Netherlands
ContactUsLinkComponent
Rat Ghrelin Receptor/GHS-R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ghrelin Receptor/GHS-R in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270973-48
Supplier:  Antibodies.com
New Product
Rat Ghrelin Receptor/GHS-R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ghrelin Receptor/GHS-R in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270973-48
Supplier:  Antibodies.com