Order Entry
Netherlands
ContactUsLinkComponent
Asymmetrical Dimethylarginine ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quanti tative determination of Asymmetrical Dimethylarginine in serum, plasma, tissue homogenates, cell lysates, cell cultur 1 * 96 Tests
Catalog # ANTIA2322-96
Supplier:  Antibodies.com
New Product
Asymmetrical Dimethylarginine ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quanti tative determination of Asymmetrical Dimethylarginine in serum, plasma, tissue homogenates, cell lysates, cell cultur 1 * 96 Tests
Catalog # ANTIA2322-96
Supplier:  Antibodies.com