Order Entry
Netherlands
ContactUsLinkComponent
Human Vitronectin/S-Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Vitronectin/S-Protein in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2163-48
Supplier:  Antibodies.com
New Product
Human Vitronectin/S-Protein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitativ e determination of human Vitronectin/S-Protein in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2163-48
Supplier:  Antibodies.com