Order Entry
Netherlands
ContactUsLinkComponent
Human Polymeric Immunoglobulin Receptor/PIGR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Polymeric Immunoglobulin Receptor/PIGR in serum, plasma, urine, saliva or ot 1 * 48 Tests
Catalog # ANTIA2173-48
Supplier:  Antibodies.com
New Product
Human Polymeric Immunoglobulin Receptor/PIGR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Polymeric Immunoglobulin Receptor/PIGR in serum, plasma, urine, saliva or ot 1 * 48 Tests
Catalog # ANTIA2173-48
Supplier:  Antibodies.com