Order Entry
Netherlands
ContactUsLinkComponent
Human Cyt 19 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cyt 19 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270655-96
Supplier:  Antibodies.com
New Product
Human Cyt 19 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Cyt 19 in serum, plasma, tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA270655-96
Supplier:  Antibodies.com