Order Entry
Netherlands
ContactUsLinkComponent
Human MEPE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MEPE in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2259-48
Supplier:  Antibodies.com
New Product
Human MEPE ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human MEPE in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2259-48
Supplier:  Antibodies.com