Order Entry
Netherlands
ContactUsLinkComponent
Human Heparanase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Heparanase 1 in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2581-48
Supplier:  Antibodies.com
New Product
Human Heparanase 1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of human Heparanase 1 in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2581-48
Supplier:  Antibodies.com