Human NAT2/AT-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human NAT2/AT-2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270745-48
Supplier: Antibodies.com