Order Entry
Netherlands
ContactUsLinkComponent
Rat Lactoferrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Lactoferrin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2095-48
Supplier:  Antibodies.com
New Product
Rat Lactoferrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of rat Lactoferrin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2095-48
Supplier:  Antibodies.com