Order Entry
Netherlands
ContactUsLinkComponent
Human ZFHX4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ZFHX4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2058-48
Supplier:  Antibodies.com
New Product
Human ZFHX4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ZFHX4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2058-48
Supplier:  Antibodies.com