Order Entry
Netherlands
ContactUsLinkComponent
27-Hydroxycholesterol ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of 27-Hydroxycholesterol in serum, plasma, cell culture supernatant, cell or tissue lysate, and other li 1 * 96 Tests
Catalog # ANTIA326318-96
Supplier:  Antibodies.com
New Product
27-Hydroxycholesterol ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative d etermination of 27-Hydroxycholesterol in serum, plasma, cell culture supernatant, cell or tissue lysate, and other li 1 * 96 Tests
Catalog # ANTIA326318-96
Supplier:  Antibodies.com