Order Entry
Netherlands
ContactUsLinkComponent
Human IGFBP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IGFBP2 in serum, plasma, urine, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2944-48
Supplier:  Antibodies.com
New Product
Human IGFBP2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IGFBP2 in serum, plasma, urine, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2944-48
Supplier:  Antibodies.com