Order Entry
Netherlands
ContactUsLinkComponent
Rat Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitr o quantitative determination of rat Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM in serum, plasma, cell culture supe 1 * 96 Tests
Catalogus # ANTIA326351-96
Supplier:  Antibodies.com
New Product
Rat Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitr o quantitative determination of rat Anti-SARS-CoV-2 Spike RBD (alpha B.1.1.7) IgM in serum, plasma, cell culture supe 1 * 96 Tests
Catalogus # ANTIA326351-96
Supplier:  Antibodies.com