Order Entry
Netherlands
ContactUsLinkComponent
Human CRF ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human CRF in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3264-48
Supplier:  Antibodies.com
New Product
Human CRF ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human CRF in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3264-48
Supplier:  Antibodies.com