Order Entry
Netherlands
ContactUsLinkComponent
Rat EGFR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat EGFR in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA2891-48
New Product
Rat EGFR ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat EGFR in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA2891-48