Order Entry
Netherlands
ContactUsLinkComponent
Human TIMP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TIMP4 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3069-48
Supplier:  Antibodies.com
New Product
Human TIMP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TIMP4 in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3069-48
Supplier:  Antibodies.com