Order Entry
Netherlands
ContactUsLinkComponent
Human ND6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ND6 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalogus # ANTIA326494-96
Leverancier:  ANTIBODIES.COM
New Product
Human ND6 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ND6 in serum, plasma, cell culture supernatant, cell or tissue lysate, and other liquid samples. 1 * 96 Tests
Catalogus # ANTIA326494-96
Leverancier:  ANTIBODIES.COM