Order Entry
Netherlands
ContactUsLinkComponent
Human IL-1F10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human IL-1F10 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA326161-96
Supplier:  ANTIBODIES.COM
New Product
Human IL-1F10 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of human IL-1F10 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA326161-96
Supplier:  ANTIBODIES.COM